Please login to your online account to display your discounted pricing

Alfa Aesar™ 5,6-Diethyl-3-(2-pyridyl)-1,2,4-triazine, tech. 90%

$65.42 - $163.16

Chemical Identifiers

Chemical Name or Material 5,6-Diethyl-3-(2-pyridyl)-1,2,4-triazine
CAS 669707-13-5
Purity Grade Technical Grade
Molecular Formula C12H14N4
Formula Weight 214.27
MDL Number MFCD09909575
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2043603

  SDS
alfa aesar™
L2043603
1g Each for $163.16

AAL20436MD

  SDS
alfa aesar™
L20436MD
250mg Each for $65.42
Description & Specifications

Specifications

Formula Weight 214.27
MDL Number MFCD09909575
Melting Point 67° to 72°C
Molecular Formula C12H14N4
Chemical Name or Material 5,6-Diethyl-3-(2-pyridyl)-1,2,4-triazine
Purity Grade Technical Grade
Assay 90%
CAS 669707-13-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.