Please login to your online account to display your discounted pricing

5,6-Dichloronicotinic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5, 6-Dichloronicotinic acid
CAS 41667-95-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H3Cl2NO2
Formula Weight 192
MDL Number MFCD00075181
Synonym 5, 6-Dichloro-3-pyridinecarboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC378790250

  SDS
thermo fisher scientific
AC378790250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 192
Physical Form Powder
Quantity 25g
Color White to beige to tan
Melting Point 163° to 168°C
CAS Max % 100.0
Water 0.5% max. (K.F.)
Assay Percent Range 98%
Chemical Name or Material 5, 6-Dichloronicotinic acid
Infrared Spectrum Authentic
CAS 41667-95-2
MDL Number MFCD00075181
CAS Min % 97.5
Synonym 5, 6-Dichloro-3-pyridinecarboxylic acid
Packaging Glass bottle
Molecular Formula C6H3Cl2NO2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.