Please login to your online account to display your discounted pricing

Alfa Aesar™ 5,6-Dichloronicotinic acid, 98+%

$97.59 - $329.41

Chemical Identifiers

Chemical Name or Material 5,6-Dichloronicotinic acid
CAS 41667-95-2
Molecular Formula C6H3Cl2NO2
Formula Weight 192
MDL Number MFCD00075181
Beilstein 383740
Synonym 5,6-Dichloropyridine-3-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2558606

 
alfa aesar™
B2558606
5g Each for $97.59

AAB2558614

 
alfa aesar™
B2558614
25g Each for $329.41
Description & Specifications

Specifications

Formula Weight 192
Chemical Name or Material 5,6-Dichloronicotinic acid
Assay ≥98%
Molecular Formula C6H3Cl2NO2
MDL Number MFCD00075181
CAS 41667-95-2
Beilstein 383740
Melting Point ≈164°C (dec.)
Synonym 5,6-Dichloropyridine-3-carboxylic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.