missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5,6-Dichloro-1H-benzo[d]imidazole-2-thiol, ≥95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals BTB02521DA

 View more versions of this product

Catalog No. BTB02521DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

5, 6-Dichloro-1H-benzo[d]imidazole-2-thiol
94.5
95%
C7H4Cl2N2S
1g
AFDOMGKBKBKUHB-UHFFFAOYSA-N
5,6-dichloro-1,3-dihydrobenzimidazole-2-thione
2774259
95%
19462-98-7
100.0
Amber glass bottle
MFCD00052398
5,6-dichloro-1h-benzo d imidazole-2-thiol, 5,6-dichlorobenzimidazole-2-thiol, 5,7dicl2sh-bzind, 2-mercapto-5,6-dichlorobenzimidazole, 5,6-dichloro-1h-benzimidazole-2-thiol, 5,6-dichloro-2-mercaptobenzimidazole, 5,6-dichloro-1h-1,3-benzodiazole-2-thiol, 2h-benzimidazole-2-thione,5,6-dichloro-1,3-dihydro, 2h-benzimidazole-2-thione, 5,6-dichloro-1,3-dihydro, 5,6-dichloro-1,3-dihydro-1,3-benzodiazole-2-thione
C1=C2C(=CC(=C1Cl)Cl)NC(=S)N2
219.083
219.09

Chemical Identifiers

19462-98-7
219.083
AFDOMGKBKBKUHB-UHFFFAOYSA-N
2774259
C1=C2C(=CC(=C1Cl)Cl)NC(=S)N2
C7H4Cl2N2S
MFCD00052398
5,6-dichloro-1h-benzo d imidazole-2-thiol, 5,6-dichlorobenzimidazole-2-thiol, 5,7dicl2sh-bzind, 2-mercapto-5,6-dichlorobenzimidazole, 5,6-dichloro-1h-benzimidazole-2-thiol, 5,6-dichloro-2-mercaptobenzimidazole, 5,6-dichloro-1h-1,3-benzodiazole-2-thiol, 2h-benzimidazole-2-thione,5,6-dichloro-1,3-dihydro, 2h-benzimidazole-2-thione, 5,6-dichloro-1,3-dihydro, 5,6-dichloro-1,3-dihydro-1,3-benzodiazole-2-thione
5,6-dichloro-1,3-dihydrobenzimidazole-2-thione
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions