Learn More
5,6-Diaminouracil sulfate, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 437660100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
5, 6-Diaminouracil sulfate | |
94.0 | |
18.80 to 21.20% C, 21.93 to 24.73% N | |
95% | |
C4H8N4O6S | |
10g | |
IKARJSDZQCSEJX-UHFFFAOYSA-N | |
5,6-diamino-1H-pyrimidine-2,4-dione;sulfuric acid | |
319643 | |
95% |
42965-55-9 | |
100.0 | |
Authentic | |
Glass bottle | |
MFCD00135845 | |
5,6-diaminopyrimidine-2,4-diol sulfate, 5,6-diaminopyrimidine-2,4-diol sulphate, diaminopyrimidine-2,4-diol; sulfuric acid, 4,5-diamino-2,6-dihydroxypyrimidine sulfate, 5,6-diaminopyrimidine-2,4 1h,3h-dione sulfate, 5,6-diaminouracil; sulfuric acid, 5?6-diaminouracil sulfate, c4h6n4o2.h2so4, 5,4-dihydroxypyrimidine sulfate, bis 5,6-diaminouracil sulphate | |
OS(O)(=O)=O.NC1=C(N)C(=O)NC(=O)N1 | |
240.19 | |
240.2 |
Chemical Identifiers
42965-55-9 | |
240.19 | |
IKARJSDZQCSEJX-UHFFFAOYSA-N | |
319643 | |
OS(O)(=O)=O.NC1=C(N)C(=O)NC(=O)N1 |
C4H8N4O6S | |
MFCD00135845 | |
5,6-diaminopyrimidine-2,4-diol sulfate, 5,6-diaminopyrimidine-2,4-diol sulphate, diaminopyrimidine-2,4-diol; sulfuric acid, 4,5-diamino-2,6-dihydroxypyrimidine sulfate, 5,6-diaminopyrimidine-2,4 1h,3h-dione sulfate, 5,6-diaminouracil; sulfuric acid, 5?6-diaminouracil sulfate, c4h6n4o2.h2so4, 5,4-dihydroxypyrimidine sulfate, bis 5,6-diaminouracil sulphate | |
5,6-diamino-1H-pyrimidine-2,4-dione;sulfuric acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash
GHS Signal Word: Warning
EINECSNumber : 256-022-8
RUO â Research Use Only