missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5,6-Diaminouracil sulfate, 95%, Thermo Scientific™

Chemical Identifiers

CAS 42965-55-9
Molecular Formula C4H8N4O6S
Molecular Weight (g/mol) 240.19
MDL Number MFCD00135845
InChI Key IKARJSDZQCSEJX-UHFFFAOYSA-N
Synonym 5,6-diaminopyrimidine-2,4-diol sulfate, 5,6-diaminopyrimidine-2,4-diol sulphate, diaminopyrimidine-2,4-diol; sulfuric acid, 4,5-diamino-2,6-dihydroxypyrimidine sulfate, 5,6-diaminopyrimidine-2,4 1h,3h-dione sulfate, 5,6-diaminouracil; sulfuric acid, 5?6-diaminouracil sulfate, c4h6n4o2.h2so4, 5,4-dihydroxypyrimidine sulfate, bis 5,6-diaminouracil sulphate
PubChem CID 319643
IUPAC Name 5,6-diamino-1,2,3,4-tetrahydropyrimidine-2,4-dione; sulfuric acid
SMILES OS(O)(=O)=O.NC1=C(N)C(=O)NC(=O)N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC437660100
SDS View Documents Promotion Details
Thermo Scientific Chemicals
437660100
10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC437660500
SDS View Documents Promotion Details
Thermo Scientific Chemicals
437660500
50g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

42965-55-9
240.19
IKARJSDZQCSEJX-UHFFFAOYSA-N
319643
OS(O)(=O)=O.NC1=C(N)C(=O)NC(=O)N1
C4H8N4O6S
MFCD00135845
5,6-diaminopyrimidine-2,4-diol sulfate, 5,6-diaminopyrimidine-2,4-diol sulphate, diaminopyrimidine-2,4-diol; sulfuric acid, 4,5-diamino-2,6-dihydroxypyrimidine sulfate, 5,6-diaminopyrimidine-2,4 1h,3h-dione sulfate, 5,6-diaminouracil; sulfuric acid, 5?6-diaminouracil sulfate, c4h6n4o2.h2so4, 5,4-dihydroxypyrimidine sulfate, bis 5,6-diaminouracil sulphate
5,6-diamino-1,2,3,4-tetrahydropyrimidine-2,4-dione; sulfuric acid

Specifications

42965-55-9
100.0
Authentic
Glass bottle
MFCD00135845
5,6-diaminopyrimidine-2,4-diol sulfate, 5,6-diaminopyrimidine-2,4-diol sulphate, diaminopyrimidine-2,4-diol; sulfuric acid, 4,5-diamino-2,6-dihydroxypyrimidine sulfate, 5,6-diaminopyrimidine-2,4 1h,3h-dione sulfate, 5,6-diaminouracil; sulfuric acid, 5?6-diaminouracil sulfate, c4h6n4o2.h2so4, 5,4-dihydroxypyrimidine sulfate, bis 5,6-diaminouracil sulphate
OS(O)(=O)=O.NC1=C(N)C(=O)NC(=O)N1
240.19
240.2
5, 6-Diaminouracil sulfate
94.0
18.80 to 21.20% C, 21.93 to 24.73% N
95%
C4H8N4O6S
10g
IKARJSDZQCSEJX-UHFFFAOYSA-N
5,6-diamino-1H-pyrimidine-2,4-dione;sulfuric acid
319643
95%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning

EINECSNumber : 256-022-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only