missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5,6,7-Trimethoxyflavone, 97%

$214.64 - $663.08

Chemical Identifiers

CAS 973-67-1
Molecular Formula C18H16O5
Molecular Weight (g/mol) 312.321
MDL Number MFCD00017457
InChI Key HJNJAUYFFFOFBW-UHFFFAOYSA-N
Synonym 5,6,7-trimethoxy-2-phenyl-4h-chromen-4-one
PubChem CID 442583
ChEBI CHEBI:2980
IUPAC Name 5,6,7-trimethoxy-2-phenylchromen-4-one
SMILES COC1=C(C(=C2C(=C1)OC(=CC2=O)C3=CC=CC=C3)OC)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2267001 View Documents Alfa Aesar™
B2267001
0.25g Each for $214.64
 
AAB2267003 View Documents Alfa Aesar™
B2267003
1g Each for $663.08
 
Specifications

Chemical Identifiers

973-67-1
312.321
HJNJAUYFFFOFBW-UHFFFAOYSA-N
442583
5,6,7-trimethoxy-2-phenylchromen-4-one
C18H16O5
MFCD00017457
5,6,7-trimethoxy-2-phenyl-4h-chromen-4-one
CHEBI:2980
COC1=C(C(=C2C(=C1)OC(=CC2=O)C3=CC=CC=C3)OC)OC

Specifications

Odorless
0.25g
C18H16O5
5,6,7-trimethoxy-2-phenyl-4h-chromen-4-one
COC1=C(C(=C2C(=C1)OC(=CC2=O)C3=CC=CC=C3)OC)OC
312.321
CHEBI:2980
97%
165°C to 167°C
973-67-1
MFCD00017457
HJNJAUYFFFOFBW-UHFFFAOYSA-N
5,6,7-trimethoxy-2-phenylchromen-4-one
442583
312.33
5,6,7-Trimethoxyflavone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures