Please login to your online account to display your discounted pricing

Alfa Aesar™ 5,6,7,8-Tetrahydro-9H-pyrido[3,2-b]indol-9-one, 98%

Chemical Identifiers

Chemical Name or Material 5,6,7,8-Tetrahydro-9H-pyrido[3,2-b]indol-9-one
CAS 198141-12-7
Formula Weight 186.21
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3527503

 
alfa aesar™
H3527503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH35275MD

 
alfa aesar™
H35275MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 186.21
Chemical Name or Material 5,6,7,8-Tetrahydro-9H-pyrido[3,2-b]indol-9-one
Physical Form Crystalline Powder
Assay 98%
CAS 198141-12-7
Color Pale brown
Melting Point 265°C (dec.)
Quantity 1g