missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5,6,7,8-Tetrahydro-2-naphthol, 98%

Manufacturer:  Alfa Aesar™ L1016414

 View more versions of this product

Catalog No. AAL1016414



Specifications

Specifications

5,6,7,8-Tetrahydro-2-naphthol
275°C to 276°C
25g
C10H12O
1101302
UMKXSOXZAXIOPJ-UHFFFAOYSA-N
5,6,7,8-tetrahydronaphthalen-2-ol
14305
148.21
>110°C (230°F)
60°C to 63°C
1125-78-6
MFCD00001738
5,6,7,8-tetrahydro-2-naphthol, 6-tetralinol, 6-hydroxytetralin, 2-naphthalenol, 5,6,7,8-tetrahydro, 5,6,7,8-tetrahydro-beta-naphthol, ac-beta-tetralol, 5,6,7,8-tetrahydro-2-hydroxynaphthalene, 2-naphthol, 5,6,7,8-tetrahydro, unii-tmr02i7n8s, 5,6,7,8-tetrahydro-2-naphthalenol
C1CCC2=C(C1)C=CC(=C2)O
148.205
CHEBI:34448
98%

Chemical Identifiers

1125-78-6
148.205
UMKXSOXZAXIOPJ-UHFFFAOYSA-N
14305
5,6,7,8-tetrahydronaphthalen-2-ol
C10H12O
MFCD00001738
5,6,7,8-tetrahydro-2-naphthol, 6-tetralinol, 6-hydroxytetralin, 2-naphthalenol, 5,6,7,8-tetrahydro, 5,6,7,8-tetrahydro-beta-naphthol, ac-beta-tetralol, 5,6,7,8-tetrahydro-2-hydroxynaphthalene, 2-naphthol, 5,6,7,8-tetrahydro, unii-tmr02i7n8s, 5,6,7,8-tetrahydro-2-naphthalenol
CHEBI:34448
C1CCC2=C(C1)C=CC(=C2)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : QL5300000

Documents