missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctene, TCI America™

Manufacturer:  TCI America T3241500MG

 View more versions of this product

Catalog No. T3241500MG



Specifications

Specifications

5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctene
110°C
53397-65-2
5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctatetraene, 5,6,11,12-Tetradehydrodibenzo[a,e][8]annulene, Dibenzo[a,e]cyclooctadien-5,11-diyne
C1=CC=C2C#CC3=CC=CC=C3C#CC2=C1
200.24
200.24
≥98.0% (HPLC)
Undesignated
500mg
C16H8
JFYLRMKUYCIWMX-UHFFFAOYSA-N
5,6,11,12-tetradehydrodibenzo[2,1-a
143044
Crystal-Powder at 20°C

Chemical Identifiers

53397-65-2
200.24
5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctatetraene, 5,6,11,12-Tetradehydrodibenzo[a,e][8]annulene, Dibenzo[a,e]cyclooctadien-5,11-diyne
5,6,11,12-tetradehydrodibenzo[2,1-a
C16H8
JFYLRMKUYCIWMX-UHFFFAOYSA-N
143044
C1=CC=C2C#CC3=CC=CC=C3C#CC2=C1
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

Documents