missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctene 98.0+%, TCI America™
Supplier: TCI America T3241100MG
Specifications
5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctene | |
110°C | |
100 mg | |
JFYLRMKUYCIWMX-UHFFFAOYSA-N | |
5,6,11,12-tetradehydrodibenzo[2,1-a | |
143044 | |
≥98.0% (HPLC) |
53397-65-2 | |
C16H8 | |
5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctatetraene, 5,6,11,12-Tetradehydrodibenzo[a,e][8]annulene, Dibenzo[a,e]cyclooctadien-5,11-diyne | |
C1=CC=C2C#CC3=CC=CC=C3C#CC2=C1 | |
200.24 | |
200.24 | |
Crystalline Powder |
Chemical Identifiers
53397-65-2 | |
200.24 | |
5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctatetraene, 5,6,11,12-Tetradehydrodibenzo[a,e][8]annulene, Dibenzo[a,e]cyclooctadien-5,11-diyne | |
5,6,11,12-tetradehydrodibenzo[2,1-a |
C16H8 | |
JFYLRMKUYCIWMX-UHFFFAOYSA-N | |
143044 | |
C1=CC=C2C#CC3=CC=CC=C3C#CC2=C1 |
Safety and Handling
TSCA : No
Recommended Storage : Freezer