Please login to your online account to display your discounted pricing

5-({[5-(Trifluoromethyl)-2-pyridyl]thio}methyl)-2-furoic acid, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  KM06169CB

 View more versions of this product

Catalog No. KM06169CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 303.26
Quantity 250mg
Assay 97%
CAS Min % 97.0
Chemical Name or Material 5-({[5-(Trifluoromethyl)-2-pyridyl]thio}methyl)-2-furoic acid
Molecular Formula C12H8F3NO3S
MDL Number MFCD00099971
CAS 238742-86-4
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Assay Percent Range 97%
Packaging Amber Glass Bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 5-({[5-(Trifluoromethyl)-2-pyridyl]thio}methyl)-2-furoic acid
CAS 238742-86-4
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H8F3NO3S
Formula Weight 303.26
MDL Number MFCD00099971
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.