Please login to your online account to display your discounted pricing

5-({[5-(Trifluoromethyl)-2-pyridyl]thio}methyl)-2-furoic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 5-({[5-(Trifluoromethyl)-2-pyridyl]thio}methyl)-2-furoic acid
CAS 238742-86-4
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H8F3NO3S
Formula Weight 303.26
MDL Number MFCD00099971
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

KM06169DA

 
fisher scientific company
KM06169DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

KM06169CB

 
fisher scientific company
KM06169CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 303.26
Quantity 1g
Assay 97%
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C12H8F3NO3S
MDL Number MFCD00099971
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Packaging Amber Glass Bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 5-({[5-(Trifluoromethyl)-2-pyridyl]thio}methyl)-2-furoic acid
CAS 238742-86-4
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.