missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5,5'-Dithiobis-(2-nitrobenzoic acid), 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC117540250

 View more versions of this product

Catalog No. AC117540250



Description

Description

Specifications

Specifications

5, 5'-Dithiobis-(2-nitrobenzoic Acid)
240.0°C to 245.0°C
69-78-3
C14H8N2O8S2
MFCD00007140
01,351
3,3'-dithiobis 6-nitrobenzoic acid, 3-carboxy-4-nitrophenyl disulfide, 5,5'-disulfanediylbis 2-nitrobenzoic acid, 5,5'-dithio-bis 2-nitrobenzoic acid, 5,5'-dithiobis 2-nitrobenzoic acid, benzoic acid, 3,3'-dithiobis 6-nitro, dithiobisnitrobenzoic acid, dithionitrobenzoic acid, dtnb, ellman's reagent
KIUMMUBSPKGMOY-UHFFFAOYSA-N
5-[(3-carboxy-4-nitrophenyl)disulfanyl]-2-nitrobenzoic acid
6254
396.34
Glass Bottle
25g
99%
[-SC6H3(NO2)CO2H]2
10, IV, 344
15, 3606
Solubility in water: freely soluble
OC(=O)C1=CC(SSC2=CC=C(C(=C2)C(O)=O)[N+]([O-])=O)=CC=C1[N+]([O-])=O
396.34
CHEBI:86228
99%

Chemical Identifiers

69-78-3
396.34
KIUMMUBSPKGMOY-UHFFFAOYSA-N
6254
5-[(3-carboxy-4-nitrophenyl)disulfanyl]-2-nitrobenzoic acid
C14H8N2O8S2
MFCD00007140
3,3'-dithiobis 6-nitrobenzoic acid, 3-carboxy-4-nitrophenyl disulfide, 5,5'-disulfanediylbis 2-nitrobenzoic acid, 5,5'-dithio-bis 2-nitrobenzoic acid, 5,5'-dithiobis 2-nitrobenzoic acid, benzoic acid, 3,3'-dithiobis 6-nitro, dithiobisnitrobenzoic acid, dithionitrobenzoic acid, dtnb, ellman's reagent
CHEBI:86228
OC(=O)C1=CC(SSC2=CC=C(C(=C2)C(O)=O)[N+]([O-])=O)=CC=C1[N+]([O-])=O
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 200-714-4

SDS Documents
Product Certifications