Please login to your online account to display your discounted pricing

5,5-Diphenylhydantoin, 99+%, ACROS Organics™

$51.88 - $51.88

Chemical Identifiers

Chemical Name or Material 5, 5-Diphenylhydantoin
Name Note 99+%
CAS 57-41-0
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0% (on dry substance)
Assay Percent Range 99+%
Molecular Formula C15H12N2O2
Formula Weight 252.27
MDL Number MFCD00005264
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC171735000

  SDS
fisher scientific acros
171735000
Plastic bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC171730050

  SDS
fisher scientific acros
171730050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC171731000

  SDS
fisher scientific acros
171731000
Plastic bottle 100g Each for $51.88
Description & Specifications

Specifications

Assay ≥99.0% (on dry substance)
Physical Form Powder
Formula Weight 252.27
Quantity 500g
Color White to almost white
Merck Index 15, 7433
Beilstein 24, 410
EINECS Number 200-328-6
RTECS Number MU1050000
CAS Max % 100.0
CAS Min % 99.0
Packaging Plastic bottle
Health Hazard 2 GHS H Statement:
Suspected of causing cancer.
Harmful if swallowed.
Suspected of damaging the unborn child.
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Synonym Phenytoin
Loss on Drying 0.5% max. (1 g, 105°C)
Chemical Name or Material 5, 5-Diphenylhydantoin
Health Hazard 1 CANCER SUSPECTED AGENT
Molecular Formula C15H12N2O2
Name Note 99+%
Assay Percent Range 99+%
MDL Number MFCD00005264
CAS 57-41-0
Safety and Handling

CANCER SUSPECTED AGENT

GHS P Statement:
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Suspected of causing cancer.
Harmful if swallowed.
Suspected of damaging the unborn child.