Please login to your online account to display your discounted pricing

5,5-Dimethyloxazolidine-2,4-dione, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5, 5-Dimethyloxazolidine-2, 4-dione
Name Note 99%
CAS 695-53-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C5H7NO3
Formula Weight 129.12
MDL Number MFCD00005379
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC204180050

  SDS
thermo fisher scientific
204180050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 129.12
Quantity 5g
Color White
Beilstein 27, 252
Merck Index 15, 3235
RTECS Number RP9100000
TSCA TSCA
Melting Point 75° to 77°C
EINECS Number 211-781-4
CAS Max % 100.0
Synonym Dimethadione
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause damage to organs through prolonged or repeated exposure.
May cause respiratory irritation.
Causes skin irritation.
CAS Min % 98.5
Appearance of Solution (5% aq. soln.) clear, colorless
MDL Number MFCD00005379
Chemical Name or Material 5, 5-Dimethyloxazolidine-2, 4-dione
Name Note 99%
CAS 695-53-4
Assay Percent Range 99%
Molecular Formula C5H7NO3