Please login to your online account to display your discounted pricing

Alfa Aesar™ 5,5-Dimethyloxazolidine-2,4-dione, 99%

$47.32 - $604.22

Chemical Identifiers

Chemical Name or Material 5,5-Dimethyloxazolidine-2,4-dione
CAS 695-53-4
Molecular Formula C5H7NO3
Formula Weight 129.12
MDL Number MFCD00005379
Beilstein 113541
Merck Index 14,3213
Synonym Dimethadione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1071403

  SDS
alfa aesar™
A1071403
1g Each for $47.32

AAA1071406

  SDS
alfa aesar™
A1071406
5g Each for $156.70

AAA1071414

  SDS
alfa aesar™
A1071414
25g Each for $604.22
Description & Specifications

Specifications

Formula Weight 129.12
Assay 99%
Quantity 1g
Molecular Formula C5H7NO3
Melting Point 77° to 80°C
Chemical Name or Material 5,5-Dimethyloxazolidine-2,4-dione
MDL Number MFCD00005379
Merck Index 14,3213
Health Hazard 2 GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
Synonym Dimethadione
RTECS Number RP9100000
Health Hazard 1 Warning
Beilstein 113541
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
CAS 695-53-4
Safety and Handling

Warning

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H361
Suspected of damaging fertility or the unborn child.