missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5,5-Dimethyl-1,3-dioxane-2-butanal, 96%

$237.05 - $237.05

Chemical Identifiers

CAS 127600-13-9
Molecular Formula C10H18O3
Formula Weight 186.25
MDL Number MFCD02683076
Synonym 5,5-Dimethyl-2-(3-formylpropyl)-1,3-dioxane; Glutaraldehyde mono-neopentyl glycol acetal
Chemical Name or Material 5,5-Dimethyl-1,3-dioxane-2-butanal
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2185809 SDS Alfa Aesar™
B2185809
10g Each for $237.05
AAB2185888 SDS Alfa Aesar™
B2185888
2.5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
It is employed as a intermediate for pharmaceuticals.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Description & Specifications

Chemical Identifiers

127600-13-9
186.25
5,5-Dimethyl-2-(3-formylpropyl)-1,3-dioxane; Glutaraldehyde mono-neopentyl glycol acetal
C10H18O3
MFCD02683076
5,5-Dimethyl-1,3-dioxane-2-butanal

Specifications

127600-13-9
186.25
10g
5,5-Dimethyl-1,3-dioxane-2-butanal
C10H18O3
MFCD02683076
5,5-Dimethyl-2-(3-formylpropyl)-1,3-dioxane; Glutaraldehyde mono-neopentyl glycol acetal
96%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning