missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5,5-Dimethyl-1,3-cyclohexanedione, 99%, ACROS Organics™
Manufacturer: Fisher Scientific Acros 116150250
Specifications
5, 5-Dimethyl-1, 3-cyclohexanedione | |
Glass bottle | |
147.0°C to 150.0°C | |
99% | |
98.5 | |
98.5% min. (HPLC) | |
(CH3)2C6H6(=O)2 | |
07, 559 | |
15, 3264 | |
Solubility in water: 5 % (25°C). Other solubilities: soluble in methanol,ethanol,benzene,chloroform,and acetic acid | |
CC1(CC(=O)CC(=O)C1)C | |
140.18 | |
140.18 | |
99% |
Authentic | |
Colorless to Yellow | |
25g | |
126-81-8 | |
100.0 | |
C8H12O2 | |
MFCD00001588 | |
01,266 | |
1,3-cyclohexanedione, 5,5-dimethyl, 5,5-dimethyl-1,3-cyclohexanedione, 5,5-dimethyldihydroresorcinol, 5,5-dimethylhydroresorcinol, cyclomethone, dimedon, dimedone, medon, methon, methone | |
BADXJIPKFRBFOT-UHFFFAOYSA-N | |
5,5-dimethylcyclohexane-1,3-dione | |
31358 | |
Liquid |
Chemical Identifiers
126-81-8 | |
140.18 | |
BADXJIPKFRBFOT-UHFFFAOYSA-N | |
31358 | |
CC1(CC(=O)CC(=O)C1)C |
C8H12O2 | |
MFCD00001588 | |
1,3-cyclohexanedione, 5,5-dimethyl, 5,5-dimethyl-1,3-cyclohexanedione, 5,5-dimethyldihydroresorcinol, 5,5-dimethylhydroresorcinol, cyclomethone, dimedon, dimedone, medon, methon, methone | |
5,5-dimethylcyclohexane-1,3-dione |
Safety and Handling
EINECSNumber : 204-804-4
RTECSNumber : GV0345000
TSCA : TSCA