Please login to your online account to display your discounted pricing

Alfa Aesar™ 5,5'-Dibromo-BAPTA, 95%

$211.03 - $649.53

Chemical Identifiers

Chemical Name or Material 5,5'-Dibromo-BAPTA
CAS 73630-11-2
Molecular Formula C22H22Br2N2O10
Formula Weight 634.23g/mol
MDL Number MFCD00036169
Synonym 1,2-Bis(2-amino-5-bromophenoxy)ethane-N,N,N',N'-tetraacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1138803

 
alfa aesar™
A11388-03
1g Each for $211.03

AAA1138806

 
alfa aesar™
A11388-06
5g Each for $649.53
Description & Specifications

Specifications

Melting Point 172° to 178°C
Quantity 1g
Chemical Name or Material 5,5'-Dibromo-BAPTA
CAS 73630-11-2
Assay 95%
Molecular Formula C22H22Br2N2O10
Formula Weight 634.23g/mol
MDL Number MFCD00036169
Synonym 1,2-Bis(2-amino-5-bromophenoxy)ethane-N,N,N',N'-tetraacetic acid
Recommended Storage 2°C to 8°C
Safety and Handling

2°C to 8°C