missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5,5'-Dibromo-2,2'-bithiophene, 99%

Manufacturer:  Alfa Aesar™ H5633714

 View more versions of this product

Catalog No. AAH5633714



Description

Description

Applications
It is an active pharmaceutical ingredient and an OLED (organic light-emitting diode) intermediate.

Solubility
Slightly soluble in water.

Notes
Recommended storage temperature: 2 - 4°C.
Specifications

Specifications

5,5'-Dibromo-2,2'-bithiophene
25g
C8H4Br2S2
5,5'-dibromo-2,2'-bithiophene, 2-bromo-5-5-bromothiophen-2-yl thiophene, 5,5-dibromo-2,2-bithiophene, 2,2'-bithiophene, 5,5'-dibromo, 5,5'-bis 2-bromothiophene, 5,5'-dibromo-2,2'bithiophene, 5,5'-dibromo-2,2-bithiophene, maybridge4_002009, acmc-209kc4, ksc491i4r
C1=C(SC(=C1)Br)C2=CC=C(S2)Br
324.048
324.06
144°C to 146°C
4805-22-5
MFCD00219110
SXNCMLQAQIGJDO-UHFFFAOYSA-N
2-bromo-5-(5-bromothiophen-2-yl)thiophene
638908
99%

Chemical Identifiers

4805-22-5
324.048
SXNCMLQAQIGJDO-UHFFFAOYSA-N
638908
C1=C(SC(=C1)Br)C2=CC=C(S2)Br
C8H4Br2S2
MFCD00219110
5,5'-dibromo-2,2'-bithiophene, 2-bromo-5-5-bromothiophen-2-yl thiophene, 5,5-dibromo-2,2-bithiophene, 2,2'-bithiophene, 5,5'-dibromo, 5,5'-bis 2-bromothiophene, 5,5'-dibromo-2,2'bithiophene, 5,5'-dibromo-2,2-bithiophene, maybridge4_002009, acmc-209kc4, ksc491i4r
2-bromo-5-(5-bromothiophen-2-yl)thiophene
Documents