Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(5-Chloro-2-thienyl)-1H-tetrazole, 97%

$370.97 - $370.97

Chemical Identifiers

Chemical Name or Material 5-(5-Chloro-2-thienyl)-1H-tetrazole
CAS 58884-89-2
Molecular Formula C5H3ClN4S
Formula Weight 186.62
MDL Number MFCD01075684
Beilstein 977762
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL15269ME

  SDS
alfa aesar™
L15269ME
500mg Each for $370.97
Description & Specifications

Specifications

Formula Weight 186.62
Chemical Name or Material 5-(5-Chloro-2-thienyl)-1H-tetrazole
Assay 97%
MDL Number MFCD01075684
CAS 58884-89-2
Beilstein 977762
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C5H3ClN4S
Melting Point ≈210°C
Quantity 500mg
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.