Please login to your online account to display your discounted pricing

5-(5-Chloro-1,2,4-thiadiazol-3-yl)thiophene-2-sulfonyl chloride, ≥95%, Maybridge

Chemical Identifiers

Chemical Name or Material 5-(5-Chloro-1, 2, 4-thiadiazol-3-yl)thiophene-2-sulfonyl chloride
CAS 306937-21-3
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C6H2Cl2N2O2S3
Formula Weight 301.2
MDL Number MFCD02180888
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPB08412CB

  SDS
fisher scientific company
SPB08412CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 301.2
Quantity 250mg
Assay 95%
CAS Min % 95.0
Packaging Amber Glass Bottle
Health Hazard 1 GHS Signal Word: Danger
Molecular Formula C6H2Cl2N2O2S3
Chemical Name or Material 5-(5-Chloro-1, 2, 4-thiadiazol-3-yl)thiophene-2-sulfonyl chloride
Assay Percent Range 95%
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
MDL Number MFCD02180888
CAS 306937-21-3
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.