Please login to your online account to display your discounted pricing

5-(5-Chloro-1,2,4-thiadiazol-3-yl)thiophene-2-sulfonyl chloride, ≥95%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SPB08412CB

 View more versions of this product

Catalog No. SPB08412CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 301.2
Quantity 250mg
Assay 95%
CAS Min % 95.0
Packaging Amber Glass Bottle
Chemical Name or Material 5-(5-Chloro-1, 2, 4-thiadiazol-3-yl)thiophene-2-sulfonyl chloride
Assay Percent Range 95%
Molecular Formula C6H2Cl2N2O2S3
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
MDL Number MFCD02180888
CAS 306937-21-3
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 5-(5-Chloro-1, 2, 4-thiadiazol-3-yl)thiophene-2-sulfonyl chloride
CAS 306937-21-3
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C6H2Cl2N2O2S3
Formula Weight 301.2
MDL Number MFCD02180888
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.