Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(5-Bromo-3-pyridyl)-1H-tetrazole, 95%

$76.50 - $76.50

Chemical Identifiers

Chemical Name or Material 5-(5-Bromo-3-pyridyl)-1H-tetrazole
CAS 211943-13-4
Molecular Formula C6H4BrN5
Formula Weight 226.04
MDL Number MFCD02093928
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL16346MD

 
alfa aesar™
L16346MD
250mg Each for $76.50
Description & Specifications

Specifications

Formula Weight 226.04
Chemical Name or Material 5-(5-Bromo-3-pyridyl)-1H-tetrazole
MDL Number MFCD02093928
CAS 211943-13-4
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Assay 95%
Molecular Formula C6H4BrN5
Melting Point 225° to 228°C
Quantity 250mg
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.