missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(5-Bromo-2-thienyl)-1H-tetrazole, 98+%

$105.32 - $325.39

Chemical Identifiers

CAS 211943-12-3
Molecular Formula C5H3BrN4S
Molecular Weight (g/mol) 231.071
MDL Number MFCD01075734
InChI Key GJHHXCMGAACACG-UHFFFAOYSA-N
Synonym 5-5-bromo-2-thienyl-1h-tetrazole, 5-5-bromothiophen-2-yl-1h-tetrazole, 1h-tetrazole, 5-5-bromo-2-thienyl, 5-5-bromothiophen-2-yl-1h-1,2,3,4-tetrazole, 5-5-bromothiophen-2-yl-2h-tetrazole, acmc-1cekq, 5-5-bromo-2-thienyl-2h-tetrazole, 5-5-bromothien-2-yl-1h-tetraazole, 2h-tetrazole,5-5-bromo-2-thienyl, 5-5-bromo-thiophen-2-yl-1h-tetrazole
PubChem CID 7021538
IUPAC Name 5-(5-bromothiophen-2-yl)-2H-tetrazole
SMILES C1=C(SC(=C1)Br)C2=NNN=N2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1634503 View Documents Alfa Aesar™
L1634503
1g Each for $325.39
 
AAL16345MD View Documents Alfa Aesar™
L16345MD
250mg Each for $105.32
 
Specifications

Chemical Identifiers

211943-12-3
231.071
GJHHXCMGAACACG-UHFFFAOYSA-N
7021538
C1=C(SC(=C1)Br)C2=NNN=N2
C5H3BrN4S
MFCD01075734
5-5-bromo-2-thienyl-1h-tetrazole, 5-5-bromothiophen-2-yl-1h-tetrazole, 1h-tetrazole, 5-5-bromo-2-thienyl, 5-5-bromothiophen-2-yl-1h-1,2,3,4-tetrazole, 5-5-bromothiophen-2-yl-2h-tetrazole, acmc-1cekq, 5-5-bromo-2-thienyl-2h-tetrazole, 5-5-bromothien-2-yl-1h-tetraazole, 2h-tetrazole,5-5-bromo-2-thienyl, 5-5-bromo-thiophen-2-yl-1h-tetrazole
5-(5-bromothiophen-2-yl)-2H-tetrazole

Specifications

~225°C
211943-12-3
MFCD01075734
GJHHXCMGAACACG-UHFFFAOYSA-N
5-(5-bromothiophen-2-yl)-2H-tetrazole
7021538
≥98%
1g
C5H3BrN4S
5-5-bromo-2-thienyl-1h-tetrazole, 5-5-bromothiophen-2-yl-1h-tetrazole, 1h-tetrazole, 5-5-bromo-2-thienyl, 5-5-bromothiophen-2-yl-1h-1,2,3,4-tetrazole, 5-5-bromothiophen-2-yl-2h-tetrazole, acmc-1cekq, 5-5-bromo-2-thienyl-2h-tetrazole, 5-5-bromothien-2-yl-1h-tetraazole, 2h-tetrazole,5-5-bromo-2-thienyl, 5-5-bromo-thiophen-2-yl-1h-tetrazole
C1=C(SC(=C1)Br)C2=NNN=N2
231.071
231.07
5-(5-Bromo-2-thienyl)-1H-tetrazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning