Please login to your online account to display your discounted pricing

(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphth-2-yl)methylamine, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC27113CB

 View more versions of this product

Catalog No. CC27113CB

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 217.35
Quantity 250mg
Assay 97%
CAS Max % 100.0
CAS 148749-58-0
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
CAS Min % 96.5
Chemical Name or Material (5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphth-2-yl)methylamine
Packaging Amber Glass Bottle
MDL Number MFCD09966143
Molecular Formula C15H23N
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material (5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphth-2-yl)methylamine
CAS 148749-58-0
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C15H23N
Formula Weight 217.35
MDL Number MFCD09966143
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents