missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphth-2-yl)methylamine, 97%, Maybridge

$71.36 - $71.36

Chemical Identifiers

CAS 148749-58-0
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C15H23N
Formula Weight 217.35
MDL Number MFCD09966143
Chemical Name or Material (5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphth-2-yl)methylamine
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC27113CB SDS Fisher Scientific Company
CC27113CB
Amber Glass Bottle 250mg Each for $71.36
CC27113DA SDS Fisher Scientific Company
CC27113DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

148749-58-0
100.0
C15H23N
MFCD09966143
97%
96.5
97%
217.35
(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphth-2-yl)methylamine

Specifications

217.35
100.0
96.5
MFCD09966143
97%
97%
250mg
148749-58-0
Amber Glass Bottle
C15H23N
(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphth-2-yl)methylamine

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger