Please login to your online account to display your discounted pricing

Alfa Aesar™ 5,5,5-Trifluoro-DL-leucine, 94%

$137.12 - $137.12

Chemical Identifiers

Chemical Name or Material 5,5,5-Trifluoro-DL-leucine
CAS 2792-72-5
Molecular Formula C6H10F3NO2
Formula Weight 185.15
MDL Number MFCD00039528
Beilstein 4664821
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL12890MC

  SDS
alfa aesar™
L12890MC
100mg Each for $137.12
Description & Specifications

Specifications

Formula Weight 185.15
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 5,5,5-Trifluoro-DL-leucine
CAS 2792-72-5
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 94%
Health Hazard 1 Warning
MDL Number MFCD00039528
Molecular Formula C6H10F3NO2
Quantity 100mg
Melting Point 273° to 275°C
Beilstein 4664821

Applications
5,5,5-Trifluoro-DL-leucine is used as an intermediate in chemical research.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Store at -80°C

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.