Please login to your online account to display your discounted pricing

5-(4-Nitrophenyl)-2-furoic acid, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC375530010

 View more versions of this product

Catalog No. AC375530010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 233.18
Quantity 1g
Molecular Formula C11H7NO5
CAS 28123-73-1
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Risk of explosion if heated under confinement.
MDL Number MFCD00094261
Synonym 5-(4-Nitrophenyl)-2-furancarboxylic acid
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 5-(4-Nitrophenyl)-2-furoic acid

Specifications

Chemical Name or Material 5-(4-Nitrophenyl)-2-furoic acid
CAS 28123-73-1
Molecular Formula C11H7NO5
Formula Weight 233.18
MDL Number MFCD00094261
Synonym 5-(4-Nitrophenyl)-2-furancarboxylic acid
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Risk of explosion if heated under confinement.

SDS & Safety Documents