Please login to your online account to display your discounted pricing

5-(4-Nitrophenyl)-2-furoic acid, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-(4-Nitrophenyl)-2-furoic acid
CAS 28123-73-1
Molecular Formula C11H7NO5
Formula Weight 233.18
MDL Number MFCD00094261
Synonym 5-(4-Nitrophenyl)-2-furancarboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC375530010

  SDS
thermo fisher scientific
AC375530010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC375530050

  SDS
thermo fisher scientific
AC375530050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 233.18
Quantity 1g
Molecular Formula C11H7NO5
CAS 28123-73-1
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Risk of explosion if heated under confinement.
MDL Number MFCD00094261
Synonym 5-(4-Nitrophenyl)-2-furancarboxylic acid
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 5-(4-Nitrophenyl)-2-furoic acid
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Risk of explosion if heated under confinement.