Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(4-Morpholinylmethyl)thiophene-2-boronic acid pinacol ester, 97%

$267.84 - $882.01

Chemical Identifiers

Chemical Name or Material 5-(4-Morpholinylmethyl)thiophene-2-boronic acid pinacol ester
CAS 950603-39-1
Molecular Formula C15H24BNO3S
Formula Weight 309.23
MDL Number MFCD09266197
Synonym 4-[5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-thienyl]methylmorpholine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2830603

  SDS
alfa aesar™
H2830603
1g Each for $267.84

AAH2830606

  SDS
alfa aesar™
H2830606
5g Each for $882.01
Description & Specifications

Specifications

Formula Weight 309.23
Chemical Name or Material 5-(4-Morpholinylmethyl)thiophene-2-boronic acid pinacol ester
Assay 97%
Molecular Formula C15H24BNO3S
MDL Number MFCD09266197
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 950603-39-1
Synonym 4-[5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-thienyl]methylmorpholine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 91° to 94°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.