Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-[4-(Methylthio)phenyl]-1H-tetrazole, 99%

$77.91 - $209.45

Chemical Identifiers

Chemical Name or Material 5-[4-(Methylthio)phenyl]-1H-tetrazole
CAS 138689-79-9
Molecular Formula C8H8N4S
Formula Weight 192.24
MDL Number MFCD03093097
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1926403

  SDS
alfa aesar™
L1926403
1g Each for $209.45

AAL19264MD

  SDS
alfa aesar™
L19264MD
250mg Each for $77.91
Description & Specifications

Specifications

Formula Weight 192.24
MDL Number MFCD03093097
CAS 138689-79-9
Assay 99%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C8H8N4S
Melting Point 214° to 215°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Quantity 1g
Chemical Name or Material 5-[4-(Methylthio)phenyl]-1H-tetrazole
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.