Please login to your online account to display your discounted pricing

5-(4-Methylphenyl)-5-phenylhydantoin, 99+%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  171430050

 View more versions of this product

Catalog No. AC171430050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Powder
Formula Weight 266.29
Quantity 5g
Color White to off-white
EINECS Number 257-028-3
Melting Point 225° to 228°C
CAS Min % 99.0
CAS Max % 100.0
Name Note 99+%
Molecular Formula C16H14N2O2
MDL Number MFCD00005263
Synonym 5-Phenyl-5-(p-tolyl)hydantoin
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material 5-(4-Methylphenyl)-5-phenylhydantoin
Assay Percent Range 99+%
CAS 51169-17-6

Specifications

Chemical Name or Material 5-(4-Methylphenyl)-5-phenylhydantoin
Name Note 99+%
CAS 51169-17-6
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C16H14N2O2
Formula Weight 266.29
MDL Number MFCD00005263
Synonym 5-Phenyl-5-(p-tolyl)hydantoin