Please login to your online account to display your discounted pricing

5-(4-Methylphenyl)-5-phenylhydantoin, 99+%, ACROS Organics™

$56.99 - $56.99

Chemical Identifiers

Chemical Name or Material 5-(4-Methylphenyl)-5-phenylhydantoin
Name Note 99+%
CAS 51169-17-6
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C16H14N2O2
Formula Weight 266.29
MDL Number MFCD00005263
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC171430010

  SDS
thermo fisher scientific
171430010
Glass bottle 1g Each for $56.99

AC171430050

  SDS
thermo fisher scientific
171430050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.0%
Formula Weight 266.29
Physical Form Powder
Quantity 1g
Color White to off-white
EINECS Number 257-028-3
Melting Point 225° to 228°C
CAS Max % 100.0
CAS Min % 99.0
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym 5-Phenyl-5-(p-tolyl)hydantoin
Name Note 99+%
Chemical Name or Material 5-(4-Methylphenyl)-5-phenylhydantoin
CAS 51169-17-6
MDL Number MFCD00005263
Assay Percent Range 99+%
Molecular Formula C16H14N2O2