Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(4-Methylphenyl)-1H-tetrazole, 98%

$98.69 - $98.69

Chemical Identifiers

Chemical Name or Material 5-(4-Methylphenyl)-1H-tetrazole
CAS 24994-04-5
Molecular Formula C8H8N4
Formula Weight 160.18
MDL Number MFCD01318164
Beilstein 131226
Synonym 5-(p-Tolyl)-1H-tetrazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1720903

  SDS
alfa aesar™
L1720903
1g Each for $98.69
Description & Specifications

Specifications

Formula Weight 160.18
CAS 24994-04-5
Chemical Name or Material 5-(4-Methylphenyl)-1H-tetrazole
MDL Number MFCD01318164
Assay 98%
Molecular Formula C8H8N4
Beilstein 131226
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 250° to 252°C
Quantity 1g
Synonym 5-(p-Tolyl)-1H-tetrazole
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.