missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(4-Methoxyphenyl)-1H-tetrazole, 98%

$155.76 - $633.81

Chemical Identifiers

CAS 6926-51-8
Molecular Formula C8H8N4O
Molecular Weight (g/mol) 176.179
MDL Number MFCD00991875
InChI Key KZGGHPBTKGLWQL-UHFFFAOYSA-N
Synonym 5-4-methoxyphenyl-2h-tetrazole, 5-4-methoxy-phenyl-2h-tetrazole, 5-4-methoxyphenyl-1h-tetrazole, 2h-tetrazole,5-4-methoxyphenyl, 5-4-methoxyphenyl-2h-tetraazole, 5-4-methoxy-phenyl-1h-tetrazole, 5-4-methoxyphenyl-1h-1,2,3,4-tetrazole, 5-4-methoxyphenyl-2h-1,2,3,4-tetrazole, 1-1h-1,2,3,4-tetraazol-5-yl-4-methoxybenzene, 4-methoxyphenyl tetrazole
PubChem CID 307902
IUPAC Name 5-(4-methoxyphenyl)-2H-tetrazole
SMILES COC1=CC=C(C=C1)C2=NNN=N2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1721003 View Documents Alfa Aesar™
L1721003
1g Each for $155.76
 
AAL1721006 View Documents Alfa Aesar™
L1721006
5g Each for $633.81
 
Specifications

Chemical Identifiers

6926-51-8
176.179
KZGGHPBTKGLWQL-UHFFFAOYSA-N
307902
COC1=CC=C(C=C1)C2=NNN=N2
C8H8N4O
MFCD00991875
5-4-methoxyphenyl-2h-tetrazole, 5-4-methoxy-phenyl-2h-tetrazole, 5-4-methoxyphenyl-1h-tetrazole, 2h-tetrazole,5-4-methoxyphenyl, 5-4-methoxyphenyl-2h-tetraazole, 5-4-methoxy-phenyl-1h-tetrazole, 5-4-methoxyphenyl-1h-1,2,3,4-tetrazole, 5-4-methoxyphenyl-2h-1,2,3,4-tetrazole, 1-1h-1,2,3,4-tetraazol-5-yl-4-methoxybenzene, 4-methoxyphenyl tetrazole
5-(4-methoxyphenyl)-2H-tetrazole

Specifications

235°C to 238°C
6926-51-8
MFCD00991875
5-4-methoxyphenyl-2h-tetrazole, 5-4-methoxy-phenyl-2h-tetrazole, 5-4-methoxyphenyl-1h-tetrazole, 2h-tetrazole,5-4-methoxyphenyl, 5-4-methoxyphenyl-2h-tetraazole, 5-4-methoxy-phenyl-1h-tetrazole, 5-4-methoxyphenyl-1h-1,2,3,4-tetrazole, 5-4-methoxyphenyl-2h-1,2,3,4-tetrazole, 1-1h-1,2,3,4-tetraazol-5-yl-4-methoxybenzene, 4-methoxyphenyl tetrazole
COC1=CC=C(C=C1)C2=NNN=N2
176.179
176.18
5-(4-Methoxyphenyl)-1H-tetrazole
1g
C8H8N4O
152619
KZGGHPBTKGLWQL-UHFFFAOYSA-N
5-(4-methoxyphenyl)-2H-tetrazole
307902
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning