Please login to your online account to display your discounted pricing

5-(4-Iodophenyl)isoxazole, 97%, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  CC12911DA

 View more versions of this product

Catalog No. CC12911DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 271.05
Quantity 1g
Assay 97%
CAS Max % 100.0
Chemical Name or Material 5-(4-Iodophenyl)isoxazole
Packaging Amber glass bottle
Assay Percent Range 97%
Molecular Formula C9H6INO
MDL Number MFCD02681898
CAS Min % 96.5
CAS 160377-48-0

Specifications

Chemical Name or Material 5-(4-Iodophenyl)isoxazole
CAS 160377-48-0
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H6INO
Formula Weight 271.05
MDL Number MFCD02681898