Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(4-Hydroxyphenyl)pentanoic acid, 98%

$63.43 - $795.57

Chemical Identifiers

Chemical Name or Material 5-(4-Hydroxyphenyl)pentanoic acid
CAS 4654-08-4
Molecular Formula C11H14O3
Formula Weight 194.23
MDL Number MFCD04039777
Synonym 5-(4-Hydroxyphenyl)valeric acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2510703

  SDS
alfa aesar™
B2510703
1g Each for $63.43

AAB2510706

  SDS
alfa aesar™
B2510706
5g Each for $213.96

AAB2510714

  SDS
alfa aesar™
B2510714
25g Each for $795.57
Description & Specifications

Specifications

Formula Weight 194.23
Molecular Formula C11H14O3
Melting Point 116° to 120°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
Synonym 5-(4-Hydroxyphenyl)valeric acid
Chemical Name or Material 5-(4-Hydroxyphenyl)pentanoic acid
MDL Number MFCD04039777
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 4654-08-4

Applications
5-(4-Hydroxyphenyl)pentanoic acid is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.