missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-(4-Hydroxyphenyl)hydantoin 98.0+%, TCI America™
Supplier: TCI America H080625G
Specifications
5-(4-Hydroxyphenyl)hydantoin | |
White-Yellow | |
MFCD00044002 | |
UMTNMIARZPDSDI-UHFFFAOYNA-N | |
5-(4-hydroxyphenyl)imidazolidine-2,4-dione | |
94233 | |
≥98.0% (N) |
2420-17-9 | |
C9H8N2O3 | |
25 g | |
OC1=CC=C(C=C1)C1NC(=O)NC1=O | |
192.17 | |
192.17 | |
Crystalline Powder |
Chemical Identifiers
2420-17-9 | |
192.17 | |
UMTNMIARZPDSDI-UHFFFAOYNA-N | |
5-(4-hydroxyphenyl)imidazolidine-2,4-dione |
C9H8N2O3 | |
MFCD00044002 | |
94233 | |
OC1=CC=C(C=C1)C1NC(=O)NC1=O |
Safety and Handling
EINECSNumber : (5)-5364
TSCA : No