Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(4-Fluorophenyl)isoxazole, 99%

$39.70 - $350.96

Chemical Identifiers

Chemical Name or Material 5-(4-Fluorophenyl)isoxazole
CAS 138716-37-7
Molecular Formula C9H6FNO
Formula Weight 163.15
MDL Number MFCD02183544
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2618803

 
alfa aesar™
H2618803
1g Each for $39.70

AAH2618814

 
alfa aesar™
H2618814
25g Each for $350.96
Description & Specifications

Specifications

Formula Weight 163.15
Chemical Name or Material 5-(4-Fluorophenyl)isoxazole
Assay 99%
Molecular Formula C9H6FNO
MDL Number MFCD02183544
CAS 138716-37-7
Melting Point 53° to 55°C
Quantity 1g