missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-(4-Fluorophenyl)cyclohexane-1,3-dione, 95%, Maybridge™

Chemical Identifiers

CAS 55579-72-1
Molecular Formula C12H11FO2
Molecular Weight (g/mol) 206.216
MDL Number MFCD00174037
InChI Key LBTLJACXBCUFEF-UHFFFAOYSA-N
Synonym 1,3-cyclohexanedione, 5-4-fluorophenyl, 1,3-cyclohexanedione,5-4-fluorophenyl, 4-3,5-dioxocyclohex-1-yl fluorobenzene, 5-4-fluoro-phenyl cyclohexane-1,3-dione, 5-4-fluoro-phenyl-cyclohexane-1,3-dione, 5-4-fluoroophenyl-1,3-cyclohexanedione, 5-4-fluorophenyl cyclohexane-1, 3-dione, 5-4-fluorophenyl cyclohexane-1,3-dione, 5-4-fluorophenyl-1,3-cyclohexanedione, acmc-20am8h
PubChem CID 730136
IUPAC Name 5-(4-fluorophenyl)cyclohexane-1,3-dione
SMILES C1C(CC(=O)CC1=O)C2=CC=C(C=C2)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CD11363DA
SDS View Documents
Fisher Scientific Company
CD11363DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CD11363EA
SDS View Documents
Fisher Scientific Company
CD11363EA
10g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

55579-72-1
206.216
LBTLJACXBCUFEF-UHFFFAOYSA-N
730136
C1C(CC(=O)CC1=O)C2=CC=C(C=C2)F
C12H11FO2
MFCD00174037
1,3-cyclohexanedione, 5-4-fluorophenyl, 1,3-cyclohexanedione,5-4-fluorophenyl, 4-3,5-dioxocyclohex-1-yl fluorobenzene, 5-4-fluoro-phenyl cyclohexane-1,3-dione, 5-4-fluoro-phenyl-cyclohexane-1,3-dione, 5-4-fluoroophenyl-1,3-cyclohexanedione, 5-4-fluorophenyl cyclohexane-1, 3-dione, 5-4-fluorophenyl cyclohexane-1,3-dione, 5-4-fluorophenyl-1,3-cyclohexanedione, acmc-20am8h
5-(4-fluorophenyl)cyclohexane-1,3-dione

Specifications

Amber glass bottle
55579-72-1
100.0
C12H11FO2
1,3-cyclohexanedione, 5-4-fluorophenyl, 1,3-cyclohexanedione,5-4-fluorophenyl, 4-3,5-dioxocyclohex-1-yl fluorobenzene, 5-4-fluoro-phenyl cyclohexane-1,3-dione, 5-4-fluoro-phenyl-cyclohexane-1,3-dione, 5-4-fluoroophenyl-1,3-cyclohexanedione, 5-4-fluorophenyl cyclohexane-1, 3-dione, 5-4-fluorophenyl cyclohexane-1,3-dione, 5-4-fluorophenyl-1,3-cyclohexanedione, acmc-20am8h
C1C(CC(=O)CC1=O)C2=CC=C(C=C2)F
206.216
206.21
5-(4-Fluorophenyl)cyclohexane-1, 3-dione
1g
94.5
95%
MFCD00174037
LBTLJACXBCUFEF-UHFFFAOYSA-N
5-(4-fluorophenyl)cyclohexane-1,3-dione
730136
95%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS