Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(4-Diethylaminobenzylidene)rhodanine, 98%

$54.55 - $215.47

Chemical Identifiers

Chemical Name or Material 5-(4-Diethylaminobenzylidene)rhodanine
CAS 35778-58-6
Molecular Formula C14H16N2OS2
Formula Weight 292.42
MDL Number MFCD00193758
Beilstein 215164
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2314506

  SDS
alfa aesar™
B2314506
5g Each for $54.55

AAB2314514

  SDS
alfa aesar™
B2314514
25g Each for $215.47
Description & Specifications

Specifications

Formula Weight 292.42
CAS 35778-58-6
Health Hazard 1 Warning
TSCA TSCA
Quantity 5g
Beilstein 215164
Molecular Formula C14H16N2OS2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00193758
Assay 98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 5-(4-Diethylaminobenzylidene)rhodanine
Melting Point 161° to 164°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.