missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(4-Cyanophenyl)-2-furoic acid, 95%

$86.76 - $345.55

Chemical Identifiers

CAS 52938-94-0
Molecular Formula C12H7NO3
Formula Weight 213.19
MDL Number MFCD06802004
Beilstein 1373676
Synonym 5-(4-Cyanophenyl)furan-2-carboxylic acid
Chemical Name or Material 5-(4-Cyanophenyl)-2-furoic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5176303 SDS Alfa Aesar™
H5176303
1g Each for $86.76
AAH5176306 SDS Alfa Aesar™
H5176306
5g Each for $345.55
Specifications Description & Specifications

Chemical Identifiers

52938-94-0
213.19
1373676
5-(4-Cyanophenyl)-2-furoic acid
C12H7NO3
MFCD06802004
5-(4-Cyanophenyl)furan-2-carboxylic acid

Specifications

1g
C12H7NO3
MFCD06802004
5-(4-Cyanophenyl)furan-2-carboxylic acid
95%
52938-94-0
213.19
1373676
5-(4-Cyanophenyl)-2-furoic acid

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger