missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(4-Chlorophenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5417603

 View more versions of this product

Catalog No. AAH5417603



Description

Description

Specifications

Specifications

5-(4-Chlorophenylcarbamoyl)-2-fluorobenzeneboronic acid
1449132-34-6
MFCD20040149
RLWSEMVWIKYUAE-UHFFFAOYSA-N
[5-[(4-chlorophenyl)carbamoyl]-2-fluorophenyl]boronic acid
73995674
97%
1g
C13H10BClFNO3
5-4-chlorophenyl carbamoyl-2-fluorophenylboronic acid, 5-4-chlorophenylcarbamoyl-2-fluorobenzeneboronic acid, 5-4-chlorophenylcarbamoyl-2-fluorophenylboronic acid
B(C1=C(C=CC(=C1)C(=O)NC2=CC=C(C=C2)Cl)F)(O)O
293.485
293.48

Chemical Identifiers

1449132-34-6
293.485
RLWSEMVWIKYUAE-UHFFFAOYSA-N
73995674
B(C1=C(C=CC(=C1)C(=O)NC2=CC=C(C=C2)Cl)F)(O)O
C13H10BClFNO3
MFCD20040149
5-4-chlorophenyl carbamoyl-2-fluorophenylboronic acid, 5-4-chlorophenylcarbamoyl-2-fluorobenzeneboronic acid, 5-4-chlorophenylcarbamoyl-2-fluorophenylboronic acid
[5-[(4-chlorophenyl)carbamoyl]-2-fluorophenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents