Please login to your online account to display your discounted pricing

5-(4-Chlorophenyl)-2-methyl-3-furoic acid, ≥97%, Maybridge

Chemical Identifiers

Chemical Name or Material 5-(4-Chlorophenyl)-2-methyl-3-furoic acid
CAS 111787-89-4
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H9ClO3
Formula Weight 236.65
MDL Number MFCD00275560
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SP00065EA

 
fisher scientific company
SP00065EA
Amber glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 236.65
Quantity 10g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 5-(4-Chlorophenyl)-2-methyl-3-furoic acid
Packaging Amber glass bottle
Assay Percent Range 97%
MDL Number MFCD00275560
CAS 111787-89-4
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Molecular Formula C12H9ClO3
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.