Please login to your online account to display your discounted pricing

5-(4-chlorophenyl)-2-furoic Acid, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-(4-Chlorophenyl)-2-furoic acid
Assay ≥95.0% (Alkalimetry)
Assay Percent Range 95+%
Molecular Formula C11H7ClO3
Formula Weight 222.63
Synonym 5-(4-Chlorophenyl)-2-furancarboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC375540050

  SDS
fisher scientific acros
AC375540050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC375540250

  SDS
thermo fisher scientific
AC375540250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC375540010

  SDS
thermo fisher scientific
AC375540010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 5-(4-Chlorophenyl)-2-furoic acid
Color Pale brown
Quantity 5g
CAS 41019-45-8
Health Hazard 1 Warning
Infrared Spectrum Authentic
Physical Form Powder
Assay ≥95.0% (Alkalimetry)
Assay Percent Range 95+%
Molecular Formula C11H7ClO3
Formula Weight 222.63
Synonym 5-(4-Chlorophenyl)-2-furancarboxylic acid
Safety and Handling

Warning