Please login to your online account to display your discounted pricing

5-(4-chlorophenyl)-2-furoic Acid, ACROS Organics™


Quantity 25g

Manufacturer: thermo fisher scientific  AC375540250

 View more versions of this product

Catalog No. AC375540250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95.0% (Alkalimetry)
Physical Form Powder
Formula Weight 222.63
Quantity 25g
Color Pale brown
CAS 41019-45-8
Assay Percent Range 95+%
Molecular Formula C11H7ClO3
Synonym 5-(4-Chlorophenyl)-2-furancarboxylic acid
Health Hazard 1 Warning
Infrared Spectrum Authentic
Chemical Name or Material 5-(4-Chlorophenyl)-2-furoic acid

Specifications

Chemical Name or Material 5-(4-Chlorophenyl)-2-furoic acid
Assay ≥95.0% (Alkalimetry)
Assay Percent Range 95+%
Molecular Formula C11H7ClO3
Formula Weight 222.63
Synonym 5-(4-Chlorophenyl)-2-furancarboxylic acid
Accessories

Warning

SDS & Safety Documents