Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(4-Chlorophenyl)-1H-tetrazole, 98+%

$79.86 - $283.27

Chemical Identifiers

Chemical Name or Material 5-(4-Chlorophenyl)-1H-tetrazole
CAS 16687-61-9
Molecular Formula C7H5ClN4
Formula Weight 180.6
MDL Number MFCD00040952
Beilstein 140053
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1173006

  SDS
alfa aesar™
L1173006
5g Each for $79.86

AAL1173014

  SDS
alfa aesar™
L1173014
25g Each for $283.27
Description & Specifications

Specifications

Formula Weight 180.6
Molecular Formula C7H5ClN4
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 5-(4-Chlorophenyl)-1H-tetrazole
MDL Number MFCD00040952
CAS 16687-61-9
Assay ≥98%
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 262° to 264°C
Quantity 5g
Beilstein 140053
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.