Please login to your online account to display your discounted pricing

5-(4-Chlorophenyl)-1-methyl-1H-imidazole-2-thiol, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 5-(4-Chlorophenyl)-1-methyl-1H-imidazole-2-thiol
Assay Percent Range 97%
Molecular Formula C10H9ClN2S
Formula Weight 224.71
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

TG00105DA

 
fisher scientific company
TG00105DA
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 224.71
Quantity 1g
Assay Percent Range 97%
CAS 17452-31-2
Chemical Name or Material 5-(4-Chlorophenyl)-1-methyl-1H-imidazole-2-thiol
Molecular Formula C10H9ClN2S
Packaging Amber Glass Bottle